Anti-TAG72 Mouse Monoclonal Antibody [clone: B72.3]
Type: Tag
Antigen: TAG72
Clonality: Monoclonal
Clone: B72.3
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
- Catalog No:
- DIABMOB288-05
- DIABMOB288
- Antigen symbol:
- TAG72
- TAG72
- Antigen name:
- Tumor-associated glycoprotein 72
- Tumor-associated glycoprotein 72
- Antibody type:
- Tag
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- B72.3
- B72.3
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa