Anti-Thyroglobulin Mouse Monoclonal Antibody [clone: DAK-Tg6]
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: Thyroglobulin
Clonality: Monoclonal
Clone: DAK-Tg6
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity:
Specifications
- Catalog No:
- DIABMOB089-05
- DIABMOB089
- Antigen symbol:
- Tg
- Tg
- Antigen name:
- Thyroglobulin
- Thyroglobulin
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- DAK-Tg6
- DAK-Tg6
- Host:
- Mouse
- Mouse
- Isotype:
- IgG2a kappa
- IgG2a kappa