Anti-T Cell Mouse Monoclonal Antibody [clone: OPD4]
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: T Cell
Clonality: Monoclonal
Clone: OPD4
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Specifications
- Catalog No:
- DIABMOB087-05
- DIABMOB087
- Antigen symbol:
- T Cell Opd4
- T Cell
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- OPD4
- OPD4
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1