Anti-S100 Mouse Monoclonal Antibody [clone: SH-B1]
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: S100
Clonality: Monoclonal
Clone: SH-B1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Specifications
- Catalog No:
- DIABMOB111-05
- DIABMOB111
- Antigen symbol:
- S100
- S100
- Antigen name:
- S100/ICaBP type calcium binding domain
- S100/ICaBP type calcium binding domain
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- SH-B1
- SH-B1
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1