Anti-RAD51 Mouse Monoclonal Antibody [clone: 51RAD01]
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: RAD51
Clonality: Monoclonal
Clone: 51RAD01
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: 0
Specifications
- Catalog No:
- DIABMOB324
- DIABMOB324-05
- Antigen symbol:
- RAD51
- RAD51
- Antigen name:
- RAD51 recombinase
- RAD51 recombinase
- Antigen synonyms:
- hRAD51|RAD51L3|DNA repair protein RAD51 homolog 1|BRCA1/BRCA2-containing complex|HRAD51|HsRad51|RecA|recombination protein A|HsRAD51|RAD51|HsT16930|subunit 5|RAD51A|E. coli|homolog of|RAD51 homolog A|RAD51D|BRCC5|RECA|RecA-like protein
- hRAD51|RAD51L3|DNA repair protein RAD51 homolog 1|BRCA1/BRCA2-containing complex|HRAD51|HsRad51|RecA|recombination protein A|HsRAD51|RAD51|HsT16930|subunit 5|RAD51A|E. coli|homolog of|RAD51 homolog A|RAD51D|BRCC5|RECA|RecA-like protein
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- 51RAD01
- 51RAD01
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa