Anti-P504S Rabbit Polyclonal Antibody
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: P504S
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: 0
Specifications
- Catalog No:
- DIABRP134-05
- DIABRP134
- Antigensymbol:
- AMACR
- AMACR
- Antikroppsnamn:
- Alpha-Methylacyl-CoA Racemase
- Alpha-Methylacyl-CoA Racemase
- Antigen synonymer:
- P504S
- P504S
- Klonalitet:
- Polyclonal
- Polyclonal
- Värd:
- Rabbit
- Rabbit