Anti-P57KIP2 Rabbit Polyclonal Antibody
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: P57KIP2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: 0
Specifications
- Catalog No:
- DIABRP104
- DIABRP104-05
- Antigen symbol:
- P57KIP2
- P57KIP2
- Antigen name:
- Cyclin-dependent kinase inhibitor 1C
- Cyclin-dependent kinase inhibitor 1C
- Antigen synonyms:
- Cyclin-dependent kinase inhibitor p57|p57Kip2|Cyclin-dependent kinase inhibitor 1C
- Cyclin-dependent kinase inhibitor p57|p57Kip2|Cyclin-dependent kinase inhibitor 1C
- Clonality:
- Polyclonal
- Polyclonal
- Host:
- Rabbit
- Rabbit
Specifications
Frequently Bought Together