Anti-p80 Anaplastic Lymphoma Kinase Mouse Monoclonal Antibody [clone: 5A4]
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: p80 Anaplastic Lymphoma Kinase
Clonality: Monoclonal
Clone: 5A4
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Specifications
- Catalog No:
- DIABMOB416-05
- DIABMOB416
- Antigen name:
- p80 Anaplastic Lymphoma Kinase
- p80 Anaplastic Lymphoma Kinase
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- 5A4
- 5A4
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1