Anti-CDKN1B Mouse Monoclonal Antibody [clone: DCS-72.F6]
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: CDKN1B
Clonality: Monoclonal
Clone: DCS-72.F6
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Specifications
- Catalog No:
- DIABMOB281
- DIABMOB281-05
- Antigen symbol:
- CDKN1B
- CDKN1B
- Antigen name:
- Cyclin-dependent kinase inhibitor 1B
- Cyclin-dependent kinase inhibitor 1B
- Antigen synonyms:
- CDKN4|KIP1|MEN1B|P27KIP1 |MEN4 |p27Kip1
- CDKN4|KIP1|MEN1B|P27KIP1 |MEN4 |p27Kip1
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- DCS-72.F6
- DCS-72.F6
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1