Anti-Myeloid Marker Mouse Monoclonal Antibody [clone: BM2]
DIABPDM084
- Antigen name:Myeloid Marker
- Antigen symbol:Myeloid Marker
- Clonality:Monoclonal
- Clone:BM2
- Host:Mouse
- Isotype:IgG1
- Epitope:? chain
- Antigen synonyms:DR alpha chain|MHC cell surface glycoprotein|HLA class II histocompatibility antigen|MHC class II antigen DRA|histocompatibility antigen HLA-DR alpha
- Size:6 mL
- Pk:6 ml
Type:
Antigen: Myeloid Marker
Clonality: Monoclonal
Clone: BM2
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: