Anti-Mesothelioma Mouse Monoclonal Antibody [clone: HBME-1]
Catalog # DIABPDM134
Supplier: DIAGNOSTIC BIOSYSTEM
Specifications
- Antigen name:Mesothelioma
- Clonality:Monoclonal
- Clone:HBME-1
- Host:Mouse
- Isotype:IgM
- Size:6 mL
- Pk:6 ml
Specifications
About this item
Type:
Antigen: Mesothelioma
Clonality: Monoclonal
Clone: HBME-1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: