Anti-CD99 Mouse Monoclonal Antibody [clone: 12 E7]
DIABPDM028
- Antigen name:CD99 molecule
- Antigen symbol:CD99
- Clonality:Monoclonal
- Clone:12 E7
- Host:Mouse
- Isotype:IgG1
- Antigen synonyms:MIC2Y|MIC2X|MSK5X|MIC2|HBA71
- Size:6 mL
- Pk:6 ml
Type:
Antigen: CD99
Clonality: Monoclonal
Clone: 12 E7
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: