Anti-Mesothelioma Mouse Monoclonal Antibody [clone: HBME-1]
Type:
Antigen: Mesothelioma
Clonality: Monoclonal
Clone: HBME-1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
- Catalog No:
- DIABMOB349
- DIABMOB349-05
- Antigen name:
- Mesothelioma
- Mesothelioma
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- HBME-1
- HBME-1
- Host:
- Mouse
- Mouse
- Isotype:
- IgM
- IgM