Anti-MCM2 Mouse Monoclonal Antibody [clone: CRCT2.1]
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: MCM2
Clonality: Monoclonal
Clone: CRCT2.1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Specifications
- Catalog No:
- DIABMOB383
- DIABMOB383-05
- Antigen symbol:
- MCM2
- MCM2
- Antigen name:
- DNA replication licensing factor MCM2
- Antigen synonyms:
- MITOTIN|minichromosome maintenance protein 2 homolog|cyclin-like 1|D3S3194|cell devision cycle-like 1|CCNL1|MCM2 minichromosome maintenance deficient 2|mitotin|minichromosome maintenance deficient 2 (mitotin)|DNA replication licensing factor MCM2|nuclear protein BM28|BM28|cdc19|CDCL1
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- CRCT2.1
- CRCT2.1
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
Specifications
Frequently Bought Together