Anti-Ki1 (CD30) Mouse Monoclonal Antibody [clone: Ber-H2]
DIABPDM018
- Antigen symbol:Ki1 (CD30)
- Clonality:Monoclonal
- Clone:Ber-H2
- Host:Mouse
- Isotype:IgG1 kappa
- Size:6 mL
- Pk:6 ml
Type:
Antigen: Ki1 (CD30)
Clonality: Monoclonal
Clone: Ber-H2
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: