Anti-KI67 Mouse Monoclonal Antibody [clone: MB67]
DIABMOB205-01
- Antigen name:Marker of proliferation Ki-67
- Antigen symbol:MKI67
- Clonality:Monoclonal
- Clone:MB67
- Host:Mouse
- Isotype:IgG1
- Size:1 mL
- Pk:1 ml
Type:
Antigen: KI67
Clonality: Monoclonal
Clone: MB67
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: