Anti-IgL Rabbit Polyclonal Antibody
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: IgL
Clonality: Polyclonal
Clone:
Conjugation:
Epitope: Light Chain
Host: Rabbit
Isotype:
Reactivity:
Specifications
- Catalog No:
- DIABRP027-05
- DIABRP027
- Antigen symbol:
- IgL
- IgL
- Antigen name:
- Immunoglobulin L
- Immunoglobulin L
- Antigen synonyms:
- lambda light chain|Ig lambda|Ig lambda light chain|Immunoglobulin lambda light chain|Igl
- lambda light chain|Ig lambda|Ig lambda light chain|Immunoglobulin lambda light chain|Igl
- Clonality:
- Polyclonal
- Polyclonal
- Host:
- Rabbit
- Rabbit
- Epitope:
- Light Chain
- Light Chain