Anti-LRP/MVP Mouse Monoclonal Antibody [clone: 1032]
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: LRP/MVP
Clonality: Monoclonal
Clone: 1032
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Specifications
- Catalog No:
- DIABMOB344-05
- DIABMOB344
- Antigen symbol:
- LRP/MVP
- LRP/MVP
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- 1032
- 1032
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1