Anti-IgL Mouse Monoclonal Antibody [clone: HP6054]
Type:
Antigen: IgL
Clonality: Monoclonal
Clone: HP6054
Conjugation:
Epitope: Light Chain
Host: Mouse
Isotype: IgG2a
Reactivity:
- Catalog No:
- DIABMOB201-05
- DIABMOB201
- Antigen symbol:
- IgL
- IgL
- Antigen name:
- Immunoglobulin L
- Immunoglobulin L
- Antigen synonyms:
- lambda light chain|Ig lambda|Ig lambda light chain|Immunoglobulin lambda light chain|Igl
- lambda light chain|Ig lambda|Ig lambda light chain|Immunoglobulin lambda light chain|Igl
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- HP6054
- HP6054
- Host:
- Mouse
- Mouse
- Isotype:
- IgG2a
- IgG2a
- Epitope:
- Light Chain
- Light Chain