Anti-Ku80 Mouse Monoclonal Antibody [clone: S10B1]
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: Ku80
Clonality: Monoclonal
Clone: S10B1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Specifications
- Catalog No:
- DIABMOB328
- DIABMOB328-05
- Antigensymbol:
- Ku80
- Ku80
- Antikroppsnamn:
- 5'-deoxyribose-5-phosphate lyase Ku80
- 5'-deoxyribose-5-phosphate lyase Ku80
- Antigen synonymer:
- complementing defective|Ku autoantigen|Ku86 autoantigen related protein 1,TLAA|CTC85|DNA repair protein XRCC5|ATP-dependent DNA helicase 2 subunit 2|86 kDa subunit of Ku antigen|80kD)|X-ray repair cross-complementing protein 5|lupus|X-ray repair|CTCBF|ATP-dependent DNA helicase II 80 kDa subunit|X-ray repair complementing defective repair in Chinese hamster cells 5 (double-strand-break rejoining|CTC box-binding factor 85 kDa subunit|repair in Chinese hamster|80kDa
- complementing defective|Ku autoantigen|Ku86 autoantigen related protein 1,TLAA|CTC85|DNA repair protein XRCC5|ATP-dependent DNA helicase 2 subunit 2|86 kDa subunit of Ku antigen|80kD)|X-ray repair cross-complementing protein 5|lupus|X-ray repair|CTCBF|ATP-dependent DNA helicase II 80 kDa subunit|X-ray repair complementing defective repair in Chinese hamster cells 5 (double-strand-break rejoining|CTC box-binding factor 85 kDa subunit|repair in Chinese hamster|80kDa
- Klonalitet:
- Monoclonal
- Monoclonal
- Klon:
- S10B1
- S10B1
- Värd:
- Mouse
- Mouse
- Isotyp:
- IgG1 kappa
- IgG1 kappa