Anti-KI67 Mouse Monoclonal Antibody [clone: MM1]
Leverantör: DIAGNOSTIC BIOSYSTEM
Certifikat
Om denna produkt
Type:
Antigen: KI67
Clonality: Monoclonal
Clone: MM1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Specifikationer
- Catalog No:
- DIABMOB237
- DIABMOB237-05
- Antigen symbol:
- MKI67
- MKI67
- Antigen name:
- Marker of proliferation Ki-67
- Marker of proliferation Ki-67
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- MM1
- MM1
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa