Anti-Involucrin Mouse Monoclonal Antibody [clone: SY5]
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: Involucrin
Clonality: Monoclonal
Clone: SY5
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Specifications
- Catalog No:
- DIABMOB270
- DIABMOB270-05
- Antigen symbol:
- IVL
- IVL
- Antigen name:
- Involucrin
- Involucrin
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- SY5
- SY5
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1