Anti-HSVI/II Rabbit Polyclonal Antibody
Katalog # DIABPDR034
Leverantör: DIAGNOSTIC BIOSYSTEM
Specifikationer
- Antigen symbol:HSVI/II
- Clonality:Polyclonal
- Host:Rabbit
- Size:6 mL
- Pk:6 ml
Specifikationer
Om denna produkt
Type:
Antigen: HSVI/II
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: