Anti-HBsAg Goat Polyclonal Antibody
Catalog # DIABPDR031
Supplier: DIAGNOSTIC BIOSYSTEM
Specifications
- Antikroppsnamn:Hepatitis B Virus Surface Antigen (ay)
- Antigensymbol:HBsAg
- Klonalitet:Polyclonal
- Värd:Goat
- Antigen synonymer:HBsAg|Hepatitis B Surface Antigen|HepB|HepB ayw|HepB surface antigen|HBVgp2
- Storlek:6 mL
- Förp.:6 ml
Specifications
About this item
Type:
Antigen: HBsAg
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: