Anti-HLA-DR Mouse Monoclonal Antibody [clone: CR3/43]
Catalog # DIABPDM082
Supplier: DIAGNOSTIC BIOSYSTEM
Specifications
- Antigen name:HLA class II histocompatibility antigen, DR alpha chain
- Antigen symbol:HLA-DR
- Clonality:Monoclonal
- Clone:CR3/43
- Host:Mouse
- Isotype:IgG1 kappa
- Epitope:? chain
- Antigen synonyms:DR alpha chain|MHC cell surface glycoprotein|HLA class II histocompatibility antigen|MHC class II antigen DRA|histocompatibility antigen HLA-DR alpha
- Size:6 mL
- Pk:6 ml
Specifications
About this item
Type:
Antigen: HLA-DR
Clonality: Monoclonal
Clone: CR3/43
Conjugation:
Epitope: ? chain
Host: Mouse
Isotype: IgG1 kappa
Reactivity: