Anti-HSVI Mouse Monoclonal Antibody [clone: 20.7.1]
DIABPDM080
- Antigen symbol:HSVI
- Clonality:Monoclonal
- Clone:20.7.1
- Host:Mouse
- Isotype:IgG1 kappa
- Size:6 mL
- Pk:6 ml
Type:
Antigen: HSVI
Clonality: Monoclonal
Clone: 20.7.1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: