Anti-HBsAg Mouse Monoclonal Antibody [clone: 3E7]
DIABPDM079
- Antigen name:Hepatitis B Virus Surface Antigen (ad/ay)
- Antigen symbol:HBsAg
- Clonality:Monoclonal
- Clone:3E7
- Host:Mouse
- Isotype:IgG1
- Antigen synonyms:HBsAg|Hepatitis B Surface Antigen|HepB|HepB ayw|HepB surface antigen|HBVgp2
- Size:6 mL
- Pk:6 ml
Type:
Antigen: HBsAg
Clonality: Monoclonal
Clone: 3E7
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: