Anti-Hairy Cell Luekemia Mouse Monoclonal Antibody [clone: DBA.44]
DIABPDM024
- Antigen name:Hairy Cell Luekemia
- Clonality:Monoclonal
- Clone:DBA.44
- Host:Mouse
- Isotype:IgM kappa
- Size:6 mL
- Pk:6 ml
Type:
Antigen: Hairy Cell Luekemia
Clonality: Monoclonal
Clone: DBA.44
Conjugation:
Epitope:
Host: Mouse
Isotype: IgM kappa
Reactivity: