Anti-IgD Rabbit Polyclonal Antibody
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: IgD
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Specifications
- Catalog No:
- DIABRP021
- DIABRP021-05
- Antigen symbol:
- IgD
- IgD
- Antigen name:
- Immunoglobulin D
- Immunoglobulin D
- Antibody type:
- Secondary
- Secondary
- Clonality:
- Polyclonal
- Polyclonal
- Host:
- Rabbit
- Rabbit