Anti-IgA Rabbit Polyclonal Antibody
Type: Secondary
Antigen: IgA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
- Catalog No:
- DIABRP020-05
- DIABRP020
- Antigen symbol:
- IgA
- IgA
- Antigen name:
- Immunoglobulin A
- Immunoglobulin A
- Antibody type:
- Secondary
- Secondary
- Clonality:
- Polyclonal
- Polyclonal
- Host:
- Rabbit
- Rabbit