Anti-HSVI Rabbit Polyclonal Antibody
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: HSVI
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Specifications
- Catalog No:
- DIABRP018-05
- DIABRP018
- Antigen symbol:
- HSVI
- HSVI
- Antigen name:
- Hepatitis B Virus Surface Antigen (ayw)
- Hepatitis B Core Antigen
- Antigen synonyms:
- HBsAg|Hepatitis B Surface Antigen|HepB|HepB ayw|HepB surface antigen|HBVgp2
- Hepatitis B virus nucleocapsid particles|HepB
- Clonality:
- Polyclonal
- Polyclonal
- Host:
- Rabbit
- Rabbit