Anti-HSP70 Rabbit Polyclonal Antibody
Type:
Antigen: HSP70
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
- Catalog No:
- DIABRP015-05
- DIABRP015
- Antigen symbol:
- HSP70
- HSP70
- Antigen name:
- Heat Shock Protein 70
- Heat Shock Protein 70
- Antigen synonyms:
- HSP70.1/HSP70.2|HSPA1|HSPA1A|HSP-70|HSPA1B|HSP70-1/HSP70-2|Heat shock 70 kDa protein 1A/1B|Heat shock 70 kDa protein 1/2|HSX70|HSP 70
- HSP70.1/HSP70.2|HSPA1|HSPA1A|HSP-70|HSPA1B|HSP70-1/HSP70-2|Heat shock 70 kDa protein 1A/1B|Heat shock 70 kDa protein 1/2|HSX70|HSP 70
- Clonality:
- Polyclonal
- Polyclonal
- Host:
- Rabbit
- Rabbit