Anti-HSP70 Mouse Monoclonal Antibody [clone: W27]
Type:
Antigen: HSP70
Clonality: Monoclonal
Clone: W27
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
- Catalog No:
- DIABMOB269-05
- DIABMOB269
- Antigen symbol:
- HSP70
- HSP70
- Antigen name:
- Heat Shock Protein 70
- Heat Shock Protein 70
- Antigen synonyms:
- HSP70.1/HSP70.2|HSPA1|HSPA1A|HSP-70|HSPA1B|HSP70-1/HSP70-2|Heat shock 70 kDa protein 1A/1B|Heat shock 70 kDa protein 1/2|HSX70|HSP 70
- HSP70.1/HSP70.2|HSPA1|HSPA1A|HSP-70|HSPA1B|HSP70-1/HSP70-2|Heat shock 70 kDa protein 1A/1B|Heat shock 70 kDa protein 1/2|HSX70|HSP 70
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- W27
- W27
- Host:
- Mouse
- Mouse
- Isotype:
- IgG2a
- IgG2a