Anti-PGR Mouse Monoclonal Antibody [clone: hpra2 + hpra3]
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: PGR
Clonality: Monoclonal
Clone: hpra2 + hpra3
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: 0
Specifications
- Catalog No:
- DIABMOB282
- DIABMOB282-05
- Antigen symbol:
- PGR
- PGR
- Antigen name:
- Progesterone Receptor
- Progesterone Receptor
- Antigen synonyms:
- PR|PRA|Nuclear receptor subfamily 3 group C member 3|Progestin receptor form B.|NR3C3|Progestin receptor form B|Progesterone receptor|PRB|Progestin receptor form A|PRGR_HUMAN|PGR
- PR|PRA|Nuclear receptor subfamily 3 group C member 3|Progestin receptor form B.|NR3C3|Progestin receptor form B|Progesterone receptor|PRB|Progestin receptor form A|PRGR_HUMAN|PGR
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- hpra2 + hpra3
- hpra2 + hpra3
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1