Order Entry
Sweden
ContactUsLinkComponent
Anti-Dff-45 Rabbit Polyclonal Antibody
Anti-Dff-45 Rabbit Polyclonal Antibody
Certificates
Anti-Dff-45 Rabbit Polyclonal Antibody

Specifications

  • Catalog No:
  • DIABRP138-05
  • DIABRP138
  • Antigen symbol:
  • Dff-45
  • DFF45
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Host:
  • Rabbit
  • Rabbit

Specifications