Anti-Cytomegalovirus Mouse Monoclonal Antibody [clone: CCH2]
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: Cytomegalovirus
Clonality: Monoclonal
Clone: CCH2
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Specifications
- Catalog No:
- DIABMOB192
- DIABMOB192-05
- Antigen symbol:
- CMV
- CMV
- Antigen name:
- Cytomegalovirus
- Cytomegalovirus
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- CCH2
- CCH2
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa