Anti-p-CK Mouse Monoclonal Antibody [clone: C-11]
Type:
Antigen: p-CK
Clonality: Monoclonal
Clone: C-11
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: 0
- Catalog No:
- DIABMOB444-05
- DIABMOB444
- Antigen symbol:
- CK Pan
- p-CK
- Antigen name:
- Cytokeratin Pan
- Pan Cytokeratin
- Antigen synonyms:
- K5|CK-5|KRT5|Type-II keratin Kb5|58 kDa cytokeratin|Keratin type II cytoskeletal 5|Keratin-5|Cytokeratin-5
- 19|wide spectrum Cytokeratin|[cytokeratins 13|P-CK|24].|Cytokeratins|17|pan-cytokeratin|14|pan CK|16|pan-CK
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- C-11
- C-11
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1