Anti-CCND3 Mouse Monoclonal Antibody [clone: DCS-28.1]
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: CCND3
Clonality: Monoclonal
Clone: DCS-28.1
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Specifications
- Catalog No:
- DIABMOB265-05
- DIABMOB265
- Antigen symbol:
- CCND3
- CCND3
- Antigen name:
- Cyclin D3
- Cyclin D3
- Antigen synonyms:
- G1/S-specific cyclin-D3,OTTHUMP00000223415|D3-type cyclin|G1/S-specific cyclin D3
- G1/S-specific cyclin-D3,OTTHUMP00000223415|D3-type cyclin|G1/S-specific cyclin D3
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- DCS-28.1
- DCS-28.1
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1