Anti-HCG Rabbit Polyclonal Antibody
Katalog # DIABPDR030
Leverantör: DIAGNOSTIC BIOSYSTEM
Specifikationer
- Antigen name:Chorionic Gonadotrophin
- Antigen symbol:HCG
- Clonality:Polyclonal
- Host:Rabbit
- Size:6 mL
- Pk:6 ml
Specifikationer
Om denna produkt
Type:
Antigen: hCG
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: