Anti-Cdx1 Rabbit Polyclonal Antibody
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: Cdx1
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Specifications
- Catalog No:
- DIABRP160-05
- DIABRP160
- Antigen symbol:
- CDX1
- CDX1
- Antigen name:
- Caudal type homeobox 1
- Caudal type homeobox 1
- Antigen synonyms:
- Homeobox protein CDX-1|Caudal-type homeobox protein 1
- Homeobox protein CDX-1|Caudal-type homeobox protein 1
- Clonality:
- Polyclonal
- Polyclonal
- Host:
- Rabbit
- Rabbit