Anti-CD68 Mouse Monoclonal Antibody [clone: KP1]
Catalog # DIABPDM066
Supplier: DIAGNOSTIC BIOSYSTEM
Specifications
- Antigen name:CD68 molecule
- Antigen symbol:CD68
- Clonality:Monoclonal
- Clone:KP1
- Host:Mouse
- Isotype:IgG1 kappa
- Antigen synonyms:macrosialin|CD68 antigen|GP110|scavenger receptor class D member 1|SCARD1|LAMP4|macrophage antigen CD68
- Size:6 mL
- Pk:6 ml
Specifications
About this item
Type:
Antigen: CD68
Clonality: Monoclonal
Clone: KP1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: