Anti-CDC47 Mouse Monoclonal Antibody [clone: 47DC141]
Type:
Antigen: CDC47
Clonality: Monoclonal
Clone: 47DC141
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
- Catalog No:
- DIABMOB340
- DIABMOB340-05
- Antigen symbol:
- CDC47
- CDC47
- Antigen name:
- Cyclin dependent kinase 47
- Cyclin dependent kinase 47
- Antigen synonyms:
- minichromosome maintenance deficient 7|CDC47 homolog|DNA replication licensing factor MCM7|homolog of S. cerevisiae Cdc47
- minichromosome maintenance deficient 7|CDC47 homolog|DNA replication licensing factor MCM7|homolog of S. cerevisiae Cdc47
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- 47DC141
- 47DC141
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1