Anti-ENG Mouse Monoclonal Antibody [clone: SN6h]
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: ENG
Clonality: Monoclonal
Clone: SN6h
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Specifications
- Catalog No:
- DIABMOB364
- DIABMOB364-05
- Antigen symbol:
- ENG
- ENG
- Antigen name:
- Endoglin
- Endoglin
- Antigen synonyms:
- Eng|AI662476|AI528660|CD105|endoglin|END|Endo|ORW1|HHT1|S-endoglin
- Eng|AI662476|AI528660|CD105|endoglin|END|Endo|ORW1|HHT1|S-endoglin
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- SN6h
- SN6h
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa