Anti-CD79a Mouse Monoclonal Antibody [clone: HM57]
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: CD79a
Clonality: Monoclonal
Clone: HM57
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Specifications
- Catalog No:
- DIABMOB118-05
- DIABMOB118
- Antigen symbol:
- CD75 (B Cell)
- CD79a
- Antigen name:
- Cluster of Differentiation 79a molecule, immunoglobulin-associated alpha
- Antigen synonyms:
- MB-1|IGA|B-cell antigen receptor complex-associated protein alpha chain|CD79a antigen immunoglobulin-associated alpha|MB-1 membrane glycoprotein
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- HM57
- HM57
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa