Order Entry
Sweden
ContactUsLinkComponent
 
Anti-CD74 B Cell Mouse Monoclonal Antibody [clone: LN2]
 
Anti-CD74 B Cell Mouse Monoclonal Antibody [clone: LN2]

 

  • Catalog No:
  • DIABMOB168
  • DIABMOB168-05
  • Antigen symbol:
  • CD74 (Lymphoid Marker)
  • CD74 B Cell
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Clone:
  • LN2
  • LN2
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa