Anti-CR2 Mouse Monoclonal Antibody [clone: 1F8]
Katalog # DIABPDM122
Supplier: DIAGNOSTIC BIOSYSTEM
Specifications
- Antikroppsnamn:Complement Component 3d receptor 2
- Antigensymbol:CR2
- Klonalitet:Monoclonal
- Klon:1F8
- Värd:Mouse
- Isotyp:IgG1 kappa
- Antigen synonymer:SLEB9|CR|CVID7|C3DRm CD21
- Storlek:6 mL
- Förp.:6 ml
Specifications
About this item
Type:
Antigen: CR2
Clonality: Monoclonal
Clone: 1F8
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: