Anti-CD15 Mouse Monoclonal Antibody [clone: MY-1]
Catalog # DIABPDM098
Supplier: DIAGNOSTIC BIOSYSTEM
Specifications
- Antigen name:3-fucosyl-N-acetyl-lactosamine
- Antigen symbol:CD15
- Clonality:Monoclonal
- Clone:MY-1
- Host:Mouse
- Isotype:IgM kappa
- Antigen synonyms:CD15|Granulocyte Associated Antigen|MC-480|BG-7|SSEA-1|SIG-3339-1000
- Size:6 mL
- Pk:6 ml
Specifications
About this item
Type:
Antigen: CD15
Clonality: Monoclonal
Clone: MY-1
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: