Anti-CD34 Mouse Monoclonal Antibody [clone: QBEND/10]
Katalog # DIABPDM050
Supplier: DIAGNOSTIC BIOSYSTEM
Specifications
- Antigen name:CD34 molecule
- Antigen symbol:CD34
- Clonality:Monoclonal
- Clone:QBEND/10
- Host:Mouse
- Isotype:IgG1 kappa
- Antigen synonyms:CD34 antigen|Mucosialin|hematopoietic progenitor cell antigen CD34|RP11-328D5.2
- Size:6 mL
- Pk:6 ml
Specifications
About this item
Type:
Antigen: CD34
Clonality: Monoclonal
Clone: QBEND/10
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: