Anti-Cd20 B Cell Rabbit Monoclonal Antibody
Type:
Antigen: Cd20 B Cell
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
- Catalog No:
- DIABRP041-05
- DIABRP041
- Antigen symbol:
- Cd20 B Cell
- CD20
- Antigen name:
- CD20 molecule
- Antigen synonyms:
- Membrane-spanning 4-domains subfamily A member 1|CVID5 MS4A2|MS4A1|Bp35|S7|LEU-16|CD20|B1
- Clonality:
- Monoclonal
- Monoclonal
- Host:
- Rabbit
- Rabbit