Anti-PTPRC Mouse Monoclonal Antibody [clone: 2B11 + PD7/26]
Supplier: DIAGNOSTIC BIOSYSTEM
Certificates
About this item
Type:
Antigen: PTPRC
Clonality: Monoclonal
Clone: 2B11 + PD7/26
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: 0
Specifications
- Catalog No:
- DIABMOB040-05
- DIABMOB040
- Antigen symbol:
- PTPRC
- PTPRC
- Antigen name:
- Protein tyrosine phosphatase receptor type C
- Protein tyrosine phosphatase receptor type C
- Antigen synonyms:
- CD45R|B220|CD45|GP180|T200|L-CA|LY5|receptor type C|LCA|protein tyrosine phosphatase
- CD45R|B220|CD45|GP180|T200|L-CA|LY5|receptor type C|LCA|protein tyrosine phosphatase
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- 2B11 + PD7/26
- 2B11 + PD7/26
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa